Learn More
Aviva Systems Biology Corp Antxr2 Antibody - N-terminal region (ARP50242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Antxr2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2310046B19Rik, AW561899, CMG-2, CMG2, cI-35ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-499