Learn More
Apexbio Technology LLC Annexin V-Cy5 Apoptosis Kit 50 assays
SDP
Catalog No. 503107121
Encompass_Preferred
Annexin V is a Ca2 -dependent phospholipid-binding protein that specifically binds phosphatidylserine (PS) PS is normally located on the inner leaflet of the cell membrane lipid bilayer but during the early stages of apoptosis it is translocated from the inner to the outer leaflet thus becoming exposed on the cell surface Annexin V exhibits high affinity for PS Cy5-labeled Annexin V can be used as a fluorescent probe allowing the detection of apoptotic cells using flow cytometry or fluorescence microscopy The staining process only takes 10 minutes
Catalog No. 50-310-7121 Supplier Apexbio Technology LLC Supplier No. K200550
Annexin V is a Ca2 -dependent phospholipid-binding protein that specifically binds phosphatidylserine (PS) PS is normally located on the inner leaflet of the cell membrane lipid bilayer but during the early stages of apoptosis it is translocated from the inner to the outer leaflet thus becoming exposed on the cell surface Annexin V exhibits high affinity for PS Cy5-labeled Annexin V can be used as a fluorescent probe allowing the detection of apoptotic cells using flow cytometry or fluorescence microscopy The staining process only takes 10 minutes