Learn More
Apexbio Technology LLC Annexin V-HF647/PI Apoptosis Kit 100assays
SDP
Catalog No. 503107173
Encompass_Preferred
Annexin V is a Ca2 -dependent phospholipid-binding protein that specifically binds to phosphatidylserine (PS) Normally PS is localized on the inner leaflet of the cell membrane lipid bilayer During the early stages of apoptosis PS translocates from the inner leaflet to the outer surface of the cell membrane exposing it to the extracellular environment Due to Annexin V s high affinity for PS fluorescently labeled Annexin V (HF647) can be used as a fluorescent probe to detect apoptotic cells through flow cytometry or fluorescence microscopy with staining taking only 10-20 minutes PI is a dye which specifically binds double-stranded DNA and emits red fluorescence under normal circumstances it cannot penetrate intact cell membranes However in late apoptotic and dead cells PI penetrates the cellular membrane staining the nuclei Dual staining with Annexin V-HF647 and PI enables clear differentiation between early apoptotic late apoptotic and dead cells
Catalog No. 50-310-7173 Supplier Apexbio Technology LLC Supplier No. K225950
Annexin V is a Ca2 -dependent phospholipid-binding protein that specifically binds to phosphatidylserine (PS) Normally PS is localized on the inner leaflet of the cell membrane lipid bilayer During the early stages of apoptosis PS translocates from the inner leaflet to the outer surface of the cell membrane exposing it to the extracellular environment Due to Annexin V s high affinity for PS fluorescently labeled Annexin V (HF647) can be used as a fluorescent probe to detect apoptotic cells through flow cytometry or fluorescence microscopy with staining taking only 10-20 minutes PI is a dye which specifically binds double-stranded DNA and emits red fluorescence under normal circumstances it cannot penetrate intact cell membranes However in late apoptotic and dead cells PI penetrates the cellular membrane staining the nuclei Dual staining with Annexin V-HF647 and PI enables clear differentiation between early apoptotic late apoptotic and dead cells