Learn More
Apexbio Technology LLC Annexin V-PE Apoptosis Kit 100tests
SDP
Catalog No. 503107220
Encompass_Preferred
Annexin V-PE Apoptosis Kit enables differentiation between early apoptotic late apoptotic and dead cells Annexin V is a Ca2 -dependent phospholipid-binding protein that specifically binds to phosphatidylserine (PS) PS is normally located on the inner leaflet of the cell membrane phospholipid bilayer However during the early stages of apoptosis PS moves from the inner surface to the outer surface of the cell membrane becoming exposed to the extracellular environment Labeled with a fluorescence dye such as PE Annexcin V is commonly used as a fluorescent probe to detect apoptosis through flow cytometry or fluorescence microscopy within 15-30 minutes Alternatively you can directly use our ready-to-use dual-staining kits Annexin V-PE/7-AAD Apoptosis Kit ( K2283) or Annexin V-PE/DAPI Apoptosis Kit (K2282)
Catalog No. 50-310-7220 Supplier Apexbio Technology LLC Supplier No. K228120
Annexin V-PE Apoptosis Kit enables differentiation between early apoptotic late apoptotic and dead cells Annexin V is a Ca2 -dependent phospholipid-binding protein that specifically binds to phosphatidylserine (PS) PS is normally located on the inner leaflet of the cell membrane phospholipid bilayer However during the early stages of apoptosis PS moves from the inner surface to the outer surface of the cell membrane becoming exposed to the extracellular environment Labeled with a fluorescence dye such as PE Annexcin V is commonly used as a fluorescent probe to detect apoptosis through flow cytometry or fluorescence microscopy within 15-30 minutes Alternatively you can directly use our ready-to-use dual-staining kits Annexin V-PE/7-AAD Apoptosis Kit ( K2283) or Annexin V-PE/DAPI Apoptosis Kit (K2282)