Learn More
Aviva Systems Biology Corp ANXA1 Antibody - N-terminal region (ARP36568_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36568_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ANXA1. Alternate names: ANX1, LPC1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-56191