Learn More
Aviva Systems Biology Corp ANXA1 Antibody - N-terminal region (ARP36569_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36569_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA1. It was validated on Western Blot. . Alternate names: ANX1, LPC1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-503