Learn More
Aviva Systems Biology Corp Anxa13 Antibody - middle region (ARP42858_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Anxa13. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1810034H17Rik, AV055219ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-038