Learn More
Aviva Systems Biology Corp ANXA2 Antibody - C-terminal region (ARP36588_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36588_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ANXA2. Alternate names: P36, ANX2, LIP2, LPC2, CAL1H, LPC2D, ANX2L4, PAP-IVApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-56202