Learn More
Aviva Systems Biology Corp ANXA4 Antibody - N-terminal region (ARP36686_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36686_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ANXA4. Alternate names: ANX4, PIG28, ZAP36ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56250