Learn More
Aviva Systems Biology Corp ANXA4 Antibody - middle region (ARP36570_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ANXA4. Alternate names: ANX4, DKFZp686H02120, MGC75105, PIG28, ZAP36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56192