Learn More
Aviva Systems Biology Corp ANXA5 Antibody - N-terminal region (ARP36687_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36687_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANXA5. Alternate names: PP4, ANX5, ENX2, RPRGL3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-56251