Learn More
Aviva Systems Biology Corp Anxa6 Antibody - C-terminal region (ARP36582_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36582_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human Anxa6. Alternate names: AW107198, Anx6, AnxVI, Cabm, Camb, ANX6, CBP68ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56197