Learn More
Aviva Systems Biology Corp Anxa6 Antibody - C-terminal region (ARP42842_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42842_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against Anxa6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse Anxa6. Alternate names: AW107198, Anx6, AnxVI, Cabm, CambApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58347