Learn More
Aviva Systems Biology Corp ANXA7 Antibody - N-terminal region (ARP36585_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36585_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANXA7. Alternate names: SNX, ANX7, SYNEXINApplicationWB. Species reactivity: Goat, Human
Catalog No. 50-014-56200