Learn More
Aviva Systems Biology Corp ANXA8L2 Antibody - middle region (ARP36574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANXA8L2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ANXA8L2. Alternate names: ANXA8, FLJ32754, bA145E20.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56194