Learn More
Stoelting™ ANY-box All Inclusive Automated System
SDP
Catalog No. 10000366 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
All inclusive automated system
Catalog No. Model
10-000-366 All inclusive automated system
1 options
Catalog No. 10-000-366 Supplier Stoelting™ Supplier No. 65000
Add to Cart
Add to Cart
Request a Quote
Stoelting™ ANY-box All Inclusive Automated System is a completely automated system designed to make behavioral tracking convenient and affordable without compromising features.
  • At the heart of ANY-box is the ANY-maze software, coupled to Stoelting's ANY-maze interface (AMi-2) device.
  • This combination of core components allows you to automate as many as four ANY-boxes per digial interface for simultaneous testing.
  • Includes:
    • ANY-box customized ANY-maze Software
    • ANY-maze digital interface
    • Universal Base Plate with Photobeam Array
    • Camera and Camera Accessories
    • Enclosures for: open field, light/dark, conditioned place preference, object recognition, hole board
    • Individual automated tests are available
    • (Note: Macintosh Operating Systems are not supported)
Specifications
For Use With (Application) Neuroscience
Model All inclusive automated system