Learn More
Stoelting™ ANY-box Behavior Test System Accessories
SDP
Catalog No. 10000361 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Auto Cond. Place Pref, Enclosure
Auto Object Recog.,Enclosure,Camera
Auto. Conditioned Place Preference
Automated Light/Dark Box
Automated Object Recognition
Automated Open Field, Clear
Automated Open Field, IR, Black
Cond. Place Pref,Photobeam Arr,4 u
Cond. Place Pref., Photobeam Array
Hole Board, Board, Enclosure
Light/Dark,Photobeam Array, 4 units
Multi-config Apparatus, IR, Camera
Novel Obj. Recognition Accessories
Open Field, Clear Photobeam Array
Open Field, Enclosure Only, Clear
Open Field, Enclosure Only, Opaque
Open Field,Enclosed, Camera, Clear
Sociability Box, Black, Enclosure
Sociability Box, Clear
Sociability Box, Clear, Enclosure
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-361 Auto Cond. Place Pref, Enclosure
10-000-360 Cond. Place Pref., Photobeam Array
10-000-363 Auto. Conditioned Place Preference
10-000-357 Automated Light/Dark Box
10-000-370 Automated Object Recognition
10-000-371 Auto Object Recog.,Enclosure,Camera
10-000-319 Automated Open Field, Clear
10-000-320 Automated Open Field, IR, Black
10-000-348 Open Field,Enclosed, Camera, Clear
10-000-347 Open Field, Clear Photobeam Array
10-000-365 Cond. Place Pref,Photobeam Arr,4 u
10-000-354 Hole Board, Board, Enclosure
10-000-359 Light/Dark,Photobeam Array, 4 units
10-000-367 Multi-config Apparatus, IR, Camera
10-000-350 Novel Obj. Recognition Accessories
10-000-317 Open Field, Enclosure Only, Clear
10-000-318 Open Field, Enclosure Only, Opaque
10-000-312 Sociability Box, Black, Enclosure
10-000-310 Sociability Box, Clear
10-000-311 Sociability Box, Clear, Enclosure
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 10-000-361 Supplier Stoelting™ Supplier No. 64001
Only null left
Add to Cart
Add to Cart
Request a Quote
Stoelting™ ANY-box Behavior Test System Accessories are completely automated systems designed to make behavioral tracking convenient and affordable without compromising features.
  • At the heart of ANY-box is the ANY-maze software, coupled to Stoelting's ANY-maze interface (AMi-2) device.
  • This combination of core components allows you to automate as many as four ANY-boxes per digial interface for simultaneous testing
Specifications
For Use With (Application) Neuroscience
Size 100 cm
Model Auto Cond. Place Pref, Enclosure