Learn More
Braintree Scientific ANY-maze Video Tracking System, ANY-maze software
SDP
Supplier:  Braintree Scientific 60000
Encompass
ANY-maze Behavioral Tracking Software - Impressively flexible, incredibly simple, intensely powerful. Track literally any animal anywhere. Intuitive, friendly design. Tracks up to 16 pieces of apparatus simultaneously. Includes more than 100 standard measures. Performs full statistical analysis of results immediately upon completion of your experiment. Comprehensive technical support. Incredibly low price. ANY-maze can automate testing in a wide variety of behavioral apparatuses. ANY-maze can perform tests in up to 16 pieces of apparatus simultaneously and has comprehensive reporting and analysis features including more than 30 built-in statistical tests. As the name implies ANY-maze can be used to track literally any animal anywhere. Using just a conventional desktop or notebook computer, and an analog, USB or FireWire camera, user can be up and running. User can also use ANY-maze to automate complex systems with multiple cameras and apparatuses.
Catalog No. 50-195-4012
 Restricted Item
Add to Cart
Request a Quote