Learn More
Bio Rad Laboratories ANYGEL STAND
Supplier:  Bio Rad Laboratories 1655131
Encompass
Stand; Bio-Rad; AnyGel; 6 rows; Holds 6 Protean gels, 12 Criterion gels, or 18 Ready Gel mini gels; includes instructions
Catalog No. 50-901-0667