Learn More
Bio Rad Laboratories ANYGEL STAND
Catalog No. 509010952
Encompass
Stand, AnyGel; Bio-Rad; Unique, convenient docking stations for any size of electrophoresis gel; Single-row; Holds 1 Protean gel, 2 Criterion gels/3 ready gel mini gels
Catalog No. 50-901-0952 Supplier Bio Rad Laboratories Supplier No. 1654131
Stand, AnyGel; Bio-Rad; Unique, convenient docking stations for any size of electrophoresis gel; Single-row; Holds 1 Protean gel, 2 Criterion gels/3 ready gel mini gels