Learn More
Sterlitech Corporation Aluminum Oxide Membrane Filters, 0.1 Micron, 13mm, 10/pk
SDP
Catalog No. 501952376
Encompass_Preferred
Aluminum oxide membranes feature uniform, densely-packed honeycomb pore structures that contribute to precise retention and high porosity characteristics.Applications:* HPLC sample and mobile phase filtration* Filtration of aggressive organic solvents* Virus concentration and analysis* Analysis of micro plastic in water* Epifluorescence microscopy* Synthesis of nanostructures (e.g. nanotubes)Features:* Fast filtration, efficient separation and high purity* High number of tightly controlled pores in honeycomb shape* Free of organic extractables and leachables* Minimum adsorption* Transparent when wet
Catalog No. 50-195-2376 Supplier Sterlitech Corporation Supplier No. 1360004
Add to Cart
Add to Cart
Aluminum oxide membranes feature uniform, densely-packed honeycomb pore structures that contribute to precise retention and high porosity characteristics.Applications:* HPLC sample and mobile phase filtration* Filtration of aggressive organic solvents* Virus concentration and analysis* Analysis of micro plastic in water* Epifluorescence microscopy* Synthesis of nanostructures (e.g. nanotubes)Features:* Fast filtration, efficient separation and high purity* High number of tightly controlled pores in honeycomb shape* Free of organic extractables and leachables* Minimum adsorption* Transparent when wet