Learn More
Sterlitech Corporation Aluminum Oxide Membrane Filters, 0.2 Micron, 25mm, 50/pk
SDP
Catalog No. 501952389
Encompass_Preferred
Aluminum oxide membranes feature uniform, densely-packed honeycomb pore structures that contribute to precise retention and high porosity characteristics.Applications:* HPLC sample and mobile phase filtration* Filtration of aggressive organic solvents* Virus concentration and analysis* Analysis of micro plastic in water* Epifluorescence microscopy* Synthesis of nanostructures (e.g. nanotubes)Features:* Fast filtration, efficient separation and high purity* High number of tightly controlled pores in honeycomb shape* Free of organic extractables and leachables* Minimum adsorption* Transparent when wet
Catalog No. 50-195-2389 Supplier Sterlitech Corporation Supplier No. 1360017
Add to Cart
Add to Cart
Aluminum oxide membranes feature uniform, densely-packed honeycomb pore structures that contribute to precise retention and high porosity characteristics.Applications:* HPLC sample and mobile phase filtration* Filtration of aggressive organic solvents* Virus concentration and analysis* Analysis of micro plastic in water* Epifluorescence microscopy* Synthesis of nanostructures (e.g. nanotubes)Features:* Fast filtration, efficient separation and high purity* High number of tightly controlled pores in honeycomb shape* Free of organic extractables and leachables* Minimum adsorption* Transparent when wet