Learn More
DENOVIX INC AO-PI VIABILITY ASSAY
Catalog No. NC2256318
Encompass
Components: 1.5 mL @ mix of 5 ug/mL AO+ 100 ug/mL PI in PBSAcridine Orange (AO) / Propidium Iodide (PI) assay are used for cell counting and viability. AO is a nucleic acid-binding fluorophore that is cell membrane permeable and suitable for selective staining of nucleated cells. PI is a nucleic acid-binding dye that is impermeable to live cells and suitable for staining dead, nucleated cells.
This item is not returnable. View return policy
Catalog No. NC2256318 Supplier DENOVIX INC Supplier No. CDAOPI1.5
Components: 1.5 mL @ mix of 5 ug/mL AO+ 100 ug/mL PI in PBSAcridine Orange (AO) / Propidium Iodide (PI) assay are used for cell counting and viability. AO is a nucleic acid-binding fluorophore that is cell membrane permeable and suitable for selective staining of nucleated cells. PI is a nucleic acid-binding dye that is impermeable to live cells and suitable for staining dead, nucleated cells.