Learn More
Nexcelom Bioscience Llc AO/PI Viability stain 5ml
Catalog No. NC0285242
Encompass
AOPI Staining Solution for the detection of live and dead nucleated mammalian cells. One 5mL vial, good for approximately 250 tests. Product warranty is valid until expiration date stated on the product label. If no expiration date is listed, warranty is valid for 12 months from the date of receipt of the product. Compatible with Cellometer Auto 2000, Cellometer Auto X4, Cellometer X2, Cellometer K2, Cellometer Vision CBA and Celigo Imaging Cytometer.
Catalog No. NC0285242 Supplier Nexcelom Bioscience Llc Supplier No. CS201065ML/DUP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
AOPI Staining Solution for the detection of live and dead nucleated mammalian cells. One 5mL vial, good for approximately 250 tests. Product warranty is valid until expiration date stated on the product label. If no expiration date is listed, warranty is valid for 12 months from the date of receipt of the product. Compatible with Cellometer Auto 2000, Cellometer Auto X4, Cellometer X2, Cellometer K2, Cellometer Vision CBA and Celigo Imaging Cytometer.