Learn More
Selleck Chemical LLC AOA hemihydrochloride 25mg
SDP
Catalog No. 502173152
Encompass_Preferred
AOA hemihydrochloride (Aminooxyacetic acid, Carboxymethoxylamine, AOAA, Aminooxyacetate hemihydrochloride) is an inhibitor ofaminobutyrate aminotransferase involved in amino acid and polyamine metabolism. AOA hemihydrochloride is also aCBS (cystathionine ?-synthase) inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3152 Supplier Selleck Chemical LLC Supplier No. S498925MG
AOA hemihydrochloride (Aminooxyacetic acid, Carboxymethoxylamine, AOAA, Aminooxyacetate hemihydrochloride) is an inhibitor ofaminobutyrate aminotransferase involved in amino acid and polyamine metabolism. AOA hemihydrochloride is also aCBS (cystathionine ?-synthase) inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.