Learn More
Apexbio Technology LLC AOA hemihydrochloride 2921-14-4 1g
SDP
Catalog No. 5000094592
Encompass_Preferred
Aminooxyacetic acid hemihydrochloride (AOA hemihydrochloride CAS 2921-14-4) is a biochemical modulator that inhibits the malate-aspartate shuttle thereby disrupting the transfer of NADH between mitochondria and the cytosol Additionally AOA suppresses -aminobutyric acid transaminase (GABA-T) leading to reduced degradation of -aminobutyric acid (GABA) This compound is widely utilized in studies of cellular energy metabolism bioenergetic processes and mechanistic investigations of GABA-related metabolic pathways in the nervous system
Catalog No. 50-000-94592 Supplier Apexbio Technology LLC Supplier No. C70581000
Aminooxyacetic acid hemihydrochloride (AOA hemihydrochloride CAS 2921-14-4) is a biochemical modulator that inhibits the malate-aspartate shuttle thereby disrupting the transfer of NADH between mitochondria and the cytosol Additionally AOA suppresses -aminobutyric acid transaminase (GABA-T) leading to reduced degradation of -aminobutyric acid (GABA) This compound is widely utilized in studies of cellular energy metabolism bioenergetic processes and mechanistic investigations of GABA-related metabolic pathways in the nervous system