Learn More
Aviva Systems Biology Corp AOC2 Antibody - middle region (ARP45055_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45055_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AOC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AOC2. Alternate names: DAO2, RAOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58963