Learn More
Cayman Chemical AOCS Canola Oil Fatty Acid Methyl Ester Standard Mixture, 1 mL
SDP
Catalog No. 502990580
Encompass_Preferred
A mixture of canola oil FAMEs; intended for use as a reference standard for the quantification of these fatty acids in canola oil by GC-FID or GC-MS
Catalog No. 50-299-0580 Supplier Cayman Chemical Supplier No. 293651 ML
A mixture of canola oil FAMEs; intended for use as a reference standard for the quantification of these fatty acids in canola oil by GC-FID or GC-MS