Learn More
Cayman Chemical AOCS LNgchaIn Faty ACD Me 50mg
SDP
Supplier:  Cayman Chemical 2866550 MG
Encompass_Preferred
A mixture of long-chain fatty acid methyl esters; contains six saturated and mono- and poly-unsaturated fatty acid methyl esters; intended for use as a reference standard for the detection of fatty acids in corn, cottonseed, soybean, safflower, sesame, poppy seed, walnut, kapok, and rice oils, among others, by GC-MS
Catalog No. 50-293-7368