Learn More
Cayman Chemical AOCS LNgchaIn Faty ACD Me 50mg
SDP
Supplier:  Cayman Chemical 2866850 MG
Encompass_Preferred
A mixture of long-chain fatty acid methyl esters; contains four saturated and mono- and poly-unsaturated fatty acid methyl esters; intended for use as a reference standard for the detection of fatty acids in olive, teaseed, and neatsfoot oils, among others, by GC-MS; supplied in an amber ampule in which the headspace has been purged with nitrogen/argon to prevent lipid oxidation
Catalog No. 50-293-9504