Learn More
Cayman Chemical AOCS Long-chain Fatty Acid Methyl Ester Standard Mixture 2, 50 mg
SDP
Catalog No. 502937069
Encompass_Preferred
A mixture of long-chain FAMEs; intended for use as a reference standard for the quantification of these long-chain fatty acids in linseed, perilla, hempseed, and rubber seed oils, among others, by GC-FID or GC-MS
Catalog No. 50-293-7069 Supplier Cayman Chemical Supplier No. 2866650 MG
A mixture of long-chain FAMEs; intended for use as a reference standard for the quantification of these long-chain fatty acids in linseed, perilla, hempseed, and rubber seed oils, among others, by GC-FID or GC-MS