Learn More
Cayman Chemical AOCS Long-chain Fatty Acid Methyl Ester Standard Mixture 3, 1 mL
SDP
Catalog No. 502990034
Encompass_Preferred
A mixture of long-chain FAMEs; intended for use as a reference standard for the quantification of these long-chain fatty acids in peanut, rapeseed, and mustard seed oils, among others, by GC-FID or GC-MS
Catalog No. 50-299-0034 Supplier Cayman Chemical Supplier No. 293661 ML
A mixture of long-chain FAMEs; intended for use as a reference standard for the quantification of these long-chain fatty acids in peanut, rapeseed, and mustard seed oils, among others, by GC-FID or GC-MS