Learn More
Cayman Chemical AOCS Medium- and Long-chain Fatty Acid Methyl Ester Standard Mixture 1, 50 mg
SDP
Catalog No. 502937349
Encompass_Preferred
A mixture of medium- and long-chain FAMEs; intended for use as a reference standard for the quantification of these fatty acids in coconut, palm kernel, babassu, and ouri-ouri oils, among others, by GC-FID or GC-MS
Catalog No. 50-293-7349 Supplier Cayman Chemical Supplier No. 2866950 MG
A mixture of medium- and long-chain FAMEs; intended for use as a reference standard for the quantification of these fatty acids in coconut, palm kernel, babassu, and ouri-ouri oils, among others, by GC-FID or GC-MS