Learn More
Cayman Chemical AOCS Medium- and Long-chain Fatty Acid Methyl Ester Standard Mixture 2, 50 mg
SDP
Catalog No. 502937369
Encompass_Preferred
A mixture of medium- and long-chain FAMEs; intended for use as a reference standard for the quantification of these medium- and long-chain fatty acids in lard, beef tallow, mutton tallow, and palm oil, among others, by GC-FID or GC-MS
Catalog No. 50-293-7369 Supplier Cayman Chemical Supplier No. 2867050 MG
A mixture of medium- and long-chain FAMEs; intended for use as a reference standard for the quantification of these medium- and long-chain fatty acids in lard, beef tallow, mutton tallow, and palm oil, among others, by GC-FID or GC-MS