Learn More
Cayman Chemical AOD-9604acetate 10mg
SDP
Catalog No. 502910047
Encompass_Preferred
A synthetic lipolytic peptide; decreases body weight, epididymal adipose tissue mass, and brown adipose tissue weight in ob/ob mice at 250 UG/kg per day; enhances cartilage regeneration in a rabbit model of collagenase-induced knee osteoarthritis
Catalog No. 50-291-0047 Supplier Cayman Chemical Supplier No. 3679010 MG
A synthetic lipolytic peptide; decreases body weight, epididymal adipose tissue mass, and brown adipose tissue weight in ob/ob mice at 250 UG/kg per day; enhances cartilage regeneration in a rabbit model of collagenase-induced knee osteoarthritis