Learn More
TARGETMOL CHEMICALS INC AOH1996 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T775205MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. AOH1996 is an orally active ligand for the replicasome component PCNA (proliferating cell nuclear antigen) targeting the transcription-replication conflict (TRC).AOH1996 interferes with the interaction of PCNA with its binding proteins leading to DNA replication stress and apoptosis.AOH1996 causes proteasome-dependent rpb1 degradation and lethal DNA damage by stabilizing the interaction between PCNA and RNA polymerase II.AOH1996 acts synergistically with DNA damaging agents to inhibit tumor cell growth. AOH1996 inhibits tumor cell growth by stabilizing the interaction between PCNA and RNA polymerase II leading to proteasome-dependent rpb1 degradation and lethal DNA damage.AOH1996 acts synergistically with DNA damaging agents. purity: 98%
Catalog No. 50-257-9140