Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AOL-Biotin Conjugate, TCI America™
SDP

Catalog No. A26591ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26591ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26591ML Supplier TCI America Supplier No. A26591ML
Add to Cart
Add to Cart

Chemical Identifiers

Synonym AOL (Aspergillus oryzae L-fucose-specific lectin)-Biotin Conjugate

Specifications

Color Yellow
Quantity 1 mL
Physical Form Liquid
Chemical Name or Material AOL-Biotin Conjugate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.