Learn More
PHYTO AB INC AOX1/2 Antibody Immunogen: AT3G22370 AT3G22360 AT3G27620 AT1G32350 AT5G64210 Background: AOX is a non-proton-pumping ubiquinol oxidase that catalyzes the reduction of oxygen to water. 150ug/EA
Supplier:  PHYTO AB INC PHY2212A
Encompass_Preferred
PhytoAB Inc. is a contract research organization and plant antibody R&D based services and products supplier. PhytoAB is privately owned company headquatered in the San Francisco Bay Area of California, USA. PhytoAB specializes in antibody production for plant scientic usage and customized services, focus on the current need of plant antibodies to assist the further investigation of the current booming results from deep genome sequencing and increasing protomics studies. We have specialized R&D and customer services teams to fulfill the needs of botany & plant sciences research and industry world wide with high quality, professional and cutting edge services, and offering complete products usage guidance and suggestions for related problem solutions.
Catalog No. NC2877300