Learn More
Waters Corp AP-1 Complete Empty Glass Column Kit, 1/pk
Supplier:  Waters Corp WAT021903
Encompass_Preferred
Waters Advanced Purification (AP) glass columns are constructed of biocompatible glass and polymeric materials for use with silica, polymer, or soft gel packings.
Catalog No. 50-785-124