Learn More
Waters Corp AP-1 Glass Insert, 10 mm x 100 mm, 1/pk
Supplier:  Waters Corp WAT021992
Encompass_Preferred
Waters Advanced Purification (AP) columns feature non-metallic construction and adjustable bed height with easy to use coarse and fine adjustments. The AP Glass columns are available in a variety of dimensions. Glass Tubes, Column Jackets, Plastic Shields and other replacement parts are sold separately.
Catalog No. 50-785-136
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.