Learn More
Medchemexpress LLC HY-133987 5mg Medchemexpress, AP-1/NF-κB activation inhibitor 1 CAS:188936-12-1 Purity:>98%
SDP
Catalog No. 502081352
Encompass_Preferred
Medchemexpress, HY-133987 5mg AP-1/NF-κB activation inhibitor 1 CAS:188936-12-1 AP-1/NF-κB activation inhibitor 1 is a potent AP-1 and NF-κB mediated transcriptional activation inhibitor (IC50=1 μM), without blocking basal transcription driven by the β-actin promoter. AP-1/NF-κB activation inhibitor 1 has a similar inhibitory effect on the production of IL-2 and IL-8 levels in stimulated cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1352 Supplier Medchemexpress LLC Supplier No. HY1339875MG
Medchemexpress, HY-133987 5mg AP-1/NF-κB activation inhibitor 1 CAS:188936-12-1 AP-1/NF-κB activation inhibitor 1 is a potent AP-1 and NF-κB mediated transcriptional activation inhibitor (IC50=1 μM), without blocking basal transcription driven by the β-actin promoter. AP-1/NF-κB activation inhibitor 1 has a similar inhibitory effect on the production of IL-2 and IL-8 levels in stimulated cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.