Learn More
TARGETMOL CHEMICALS INC AP-1/NF-KAPPAB ACTIVATIO 5MG
SDP
Catalog No. 502578979
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AP-1/NF-(kappa)B activation inhibitor 1 is a potent inhibitor of AP-1 and NF-(kappa)B mediated transcriptional activation ( IC 50 =1 uM) does not blocking basal transcription driven by the (beta)-actin promoter. AP-1/NF-(kappa)B activation inhibitor 1 has a similar inhibitory effect on the production of IL-2 and IL-8 levels in stimulated cells [1]. purity: 99%
Catalog No. 50-257-8979 Supplier TARGETMOL CHEMICALS INC Supplier No. T96565MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AP-1/NF-(kappa)B activation inhibitor 1 is a potent inhibitor of AP-1 and NF-(kappa)B mediated transcriptional activation ( IC 50 =1 uM) does not blocking basal transcription driven by the (beta)-actin promoter. AP-1/NF-(kappa)B activation inhibitor 1 has a similar inhibitory effect on the production of IL-2 and IL-8 levels in stimulated cells [1]. purity: 99%