Learn More
Waters Corp AP-1 Plastic Shield, 10 mm x 600 mm, 1/pk
Catalog No. 50785133 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Advanced Purification (AP) columns feature non-metallic construction and adjustable bed height with easy to use coarse and fine adjustments. The AP Glass columns are available in a variety of dimensions. Glass Tubes, Column Jackets, Plastic Shields and other replacement parts are sold separately.
Catalog No. 50-785-133 Supplier Waters Corp Supplier No. WAT021947
Waters Advanced Purification (AP) columns feature non-metallic construction and adjustable bed height with easy to use coarse and fine adjustments. The AP Glass columns are available in a variety of dimensions. Glass Tubes, Column Jackets, Plastic Shields and other replacement parts are sold separately.