Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AP-2 gamma/TFAP2C protein, human (His) | >95.0% | 100 UG
SDP

Catalog No. 5000293764
Encompass_Preferred

Recombinant human AP-2 gamma/TFAP2C protein fragment (residues L128-V223) expressed in E. coli with an N-terminal 10xHis tag. Supplied as a sterile, lyophilized powder from PBS (pH 7.4), the product is >95% pure and intended for biochemical and DNA-binding assays. Store frozen and follow recommended reconstitution and aliquoting practices for stability.

  • Residues L128-V223 representing the human TFAP2C fragment.
  • N-terminal 10xHis tag for affinity purification.
  • Expression system: E. coli.
  • Molecular weight approximately 10-14 kDa.
  • Purity greater than 95.0% by reducing SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Recommended reconstitution concentration not less than 100 μg/mL.
  • Storage: -20°C up to 2 years; after reconstitution stable at 4°C for 1 week.
  • Available in multiple sizes, including 100 μg.

Catalog No. 50-002-93764 Supplier Medchemexpress LLC Supplier No. HYP76728100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human AP-2 gamma/TFAP2C protein fragment (residues L128-V223) expressed in E. coli with an N-terminal 10xHis tag. Supplied as a sterile, lyophilized powder from PBS (pH 7.4), the product is >95% pure and intended for biochemical and DNA-binding assays. Store frozen and follow recommended reconstitution and aliquoting practices for stability.

  • Residues L128-V223 representing the human TFAP2C fragment.
  • N-terminal 10xHis tag for affinity purification.
  • Expression system: E. coli.
  • Molecular weight approximately 10-14 kDa.
  • Purity greater than 95.0% by reducing SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Recommended reconstitution concentration not less than 100 μg/mL.
  • Storage: -20°C up to 2 years; after reconstitution stable at 4°C for 1 week.
  • Available in multiple sizes, including 100 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.