Learn More
TARGETMOL CHEMICALS INC AP-C5 5MG
SDP
Catalog No. 502579614
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AP-C5 is a compound with selective inhibition of guanosine 3'5' cyclic monophosphate (cGMP)-dependent protein kinase II (cGKII) with a Pic50 value of 7.2 for cGMP and can be used in the study of diarrheal diseases. purity: 99%
Catalog No. 50-257-9614 Supplier TARGETMOL CHEMICALS INC Supplier No. T605005MG
Also available in 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AP-C5 is a compound with selective inhibition of guanosine 3'5' cyclic monophosphate (cGMP)-dependent protein kinase II (cGKII) with a Pic50 value of 7.2 for cGMP and can be used in the study of diarrheal diseases. purity: 99%