Learn More
Seracare Life Sciences Inc AP CHEMI NA BLOTTING KIT
Supplier:  Seracare Life Sciences Inc 59100029
Encompass_Preferred
The Detector AP Chemiluminescent Blotting Kit is are designed for the hybridization and chemiluminescent detection of biotinylated DNA or RNA probes on membranes, providing the highest sensitivity of KPL's non-radioactive nucleic acid detection systems. Ideal for detection of Northern blots or single copy genomic Southern blots, moderately expressed transcripts in just 50 ng of total RNA and single copy genes in 1-5 ug human genomic DNA can be observed in as little as 10 minutes. The kit contains all reagents necessary to perform hybridization and detection of membrane-bound nucleic acids, except for the biointylated probe. Applications include Southern blotting, Northern blotting, bacterial colony hyridizations, plaque hybridizations, and dot blotting. Key benefits include detection of transcripts in 50 ng of total RNA or single copy genes in 1-5 ug genomic DNA in as little as 10 minutes, chemiluminescent light emission lasts for up to 5 days, and blots may be stripped and reprobed.
Catalog No. 50-675-07
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.