Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ AP Chemistry Determination of the Molecular Mass of Liquid
Shop All Aldon ProductsDescription
- How the kit works:
- In this experiment, a volatile liquid is placed in a test tube sealed with a stopper containing a small hole and heated in a hot water bath until it vaporizes, allowing excess vapor to escape. The tube is then rapidly cooled in an ice bath to condense the vapor, after which the mass of the tube and condensed liquid is determined
- Kit Includes:
- 2 x 25 mL Methyl Alcohol
- 2 x 25 mL Isopropyl Alcohol
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Class Size | 15 Lab Groups |
| Product Type | AP Chemistry Lab Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?