Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ AP Chemistry Determination of the Properties of Buffer Solutions
Shop All Aldon ProductsDescription
- How the kit works:
- In this experiment, three buffer solutions with different pH values are prepared to demonstrate their resistance to pH changes when small amounts of acids or bases are added. The behavior of these buffers is then compared with water and a non-buffer solution (NaCl), which exhibit much larger pH changes under the same conditions
- Kit Includes:
- 1 x 30 g Sodium Acetate, trihydrate
- 2 x 25 g Ammonium Chloride
- 1 x 30 g Sodium Bicarbonate
- 4 x 25 g Sodium Chloride
- 1 x 23 mL Acetic Acid EZ-Prep, to make 800 mL of 0.5 M Solution
- 1 x 27 mL Ammonium Hydroxide, EZ-Prep, to make 800 mL of 0.5 M Solution
- 1 x 15 mL Sodium Hydroxide, 1.0 N Solution
- 1 x 15 mL Hydrochloric Acid, 1.0 N Solution
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Class Size | 15 Lab Groups |
| Product Type | AP Chemistry Lab Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?