Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Chemistry Dissociation Constants of Weak Acids

Catalog No. S001292 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size Grade
S001292 15 Lab Groups 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.
Catalog No. S001292 Supplier Innovating Science™ Supplier No. IS8008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Phenolphthalein, which is known to the State of California to cause cancer. For more information go to www.P65Warnings.ca.gov.

Explore the principles of equilibrium. An essential hands-on lab for mastering acid-base chemistry and experimental analysis.

  • How the kit works:
    • Students determine the equilibrium constant of three unknown acids and compare the data to known values to identify them
  • Kit Includes:
    • 1 x 10 g Benzoic Acid Crystals
    • 1 x 10 g Potassium Hydrogen Phthalate
    • 1 x 10 g Sodium Bisulfate (Sodium Hydrogen Sulfate)
    • 10 x 250 mL Sodium Hydroxide, 0.1 M Solution
    • 4 x 25 mL Phenolphthalein, 1% in IPA Solution

Specifications

Grade Level High School, Higher Ed
Grade 9 to 12
Class Size 15 Lab Groups
Product Type AP Chemistry Lab Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.