Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ AP Chemistry Freezing Point Depression to Determine Molecular Mass
Shop All Aldon ProductsDescription
- How the kit works:
- Molecular mass can help identify unknown compounds. In this experiment, it is determined by measuring freezing point depression, a colligative property that depends on the number of dissolved particles
- Para-dichlorobenzene is dissolved in the nonpolar solvent 2,6-di-tert-butyl-4-methylphenol, which has a melting point of approximately 70°C, and the resulting temperature change is measured. The solvent's depression constant is then calculated, and the procedure is repeated with two unknown compounds to determine their molecular weights
- Kit Includes:
- 2 x 25 g Di-Tert-Butyl-4-Methylphenol Crystals
- 1 x 25 g p-Dichlorobenzene Crystals
- 1 x 25 g Stearic Acid Flakes
- 1 x 25 g Naphthalene Flakes
Specifications
Specifications
| Grade Level | High School, Higher Ed |
| Grade | 9 to 12 |
| Class Size | 15 Lab Groups |
| Product Type | AP Chemistry Lab Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?